EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Omeprazole limited sampling strategies to predict area under the concentration-time curve ratios: implications for cytochrome P450 2C19 and 3A phenotyping.
PMID 22009190
Abstract
To develop a limited sampling strategy (LSS) to predict area under the concentration-time curve (AUC) ratios of omeprazole (AUC(OPZ)) to its metabolites 5-hydroxyomeprazole (AUC(5OH)) and omeprazole sulfone (AUC(SUL)) as phenotyping parameters for cytochrome P450 (CYP) 2C19 and 3A. Data were obtained from 37 (4 women) Caucasian, Chinese, and Korean healthy adults from three published studies. The AUC(OPZ), AUC(5OH), and AUC(SUL) were calculated via noncompartmental analysis. Observed AUC(OPZ, OBS)/AUC(5OH, OBS) and AUC(OPZ, OBS)/AUC(SUL, OBS) were determined. Plasma concentrations of omeprazole, 5-hydroxyomeprazole, and omeprazole sulfone at 1, 1.5, 2, 3, 4, 6, and 8 h post-dose were used to generate limited sampling strategy (LSS) models to predict AUC(OPZ,PRE)/AUC(5OH,PRE) and AUC(OPZ,PRE/)AUC(SUL,PRE). Bias and precision were assessed via percentage mean prediction error (%MPE) and percentage mean absolute error (%MAE), with acceptable limits being <15%. For CYP2C19, the AUC(OPZ,OBS)/AUC(5OH,OBS) was [mean ± standard deviation (SD)] 2.10 ± 1.63. Five LSS models of AUC(OPZ,PRE)/AUC(5OH,PRE) were generated, but none met the bias or precision criteria. Upon stratification by CYP2C19 genotype and ethnicity, a three-timepoint (at 1, 2, and 4 h) LSS model accurately predicted AUC(OPZ)/AUC(5OH) in Caucasian CYP2C19*1/*1 subjects. For CYP3A, AUC(OPZ,OBS)/AUC(SUL,OBS) (mean ± SD) was 1.79 ± 0.67. All LSS models had unacceptable %MAE, even when stratified by CYP2C19 genotype and ethnicity. A LSS model to predict AUC(OPZ)/AUC(5OH), and thus CYP2C19 activity, was generated for Caucasian CYP2C19*1/*1 subjects. However, additional model validation is needed prior to general use. LSS models to predict AUC(OPZ)/AUC(SUL), and thus CYP3A activity, were not possible, even upon stratification by CYP2C19 genotype and ethnicity.
Related Materials