EMAIL THIS PAGE TO A FRIEND
MMWR. Morbidity and mortality weekly report
Carbapenem-resistant Klebsiella pneumoniae associated with a long-term--care facility --- West Virginia, 2009-2011.
PMID 22012114
Abstract
On January 27, 2011, a West Virginia county health department was notified of a cluster of carbapenem-resistant Klebsiella pneumoniae (CRKP) cases detected by a local hospital (hospital A). CRKP infections frequently are resistant to a majority of antimicrobial agents and have an increased risk for morbidity and mortality. The West Virginia Bureau for Public Health (WVBPH) conducted field investigations to identify all cases, characterize risk factors for infection, and abstract data for a matched case-control study. Nineteen case-patients and 38 control patients were identified. Infection with CRKP was associated with admission from or prior stay at a local long-term--care facility (LTCF A). Pulsed-field gel electrophoresis (PFGE) analysis indicated that all five hospital A clinical specimens and all 11 point prevalence survey isolates from LTCF A were closely related. This is the first outbreak of CRKP identified in West Virginia. Recommendations to LTCF A included the following: 1) initiate surveillance for multidrug resistant organisms; 2) revise and improve infection prevention and control activities within the facility; 3) educate residents and their families, physicians, and staff members about CRKP; and 4) identify qualified personnel to coordinate infection control functions within the facility. Although LTCF A has made significant improvements, the outbreak investigation is ongoing. Additional site visits have been conducted, and additional colonized residents have been identified; the last clinical case was detected in July. These findings demonstrate the interconnectedness of the health-care system and factors potentially contributing to transmission of infection. Interventions targeting all levels of care are needed to prevent further CRKP transmission.