EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Microfluidic tool based on the antibody-modified paramagnetic particles for detection of 8-hydroxy-2'-deoxyguanosine in urine of prostate cancer patients.
PMID 22012838
Abstract
Guanosine derivatives are important for diagnosis of oxidative DNA damage including 8-hydroxy-2'-deoxyguanosine (8-OHdG) as one of the most abundant products of DNA oxidation. This compound is commonly determined in urine, which makes 8-OHdG a good non-invasive marker of oxidation stress. In this study, we optimized and tested the isolation of 8-OHdG from biological matrix by using paramagnetic particles with an antibody-modified surface. 8-OHdG was determined using 1-naphthol generated by alkaline phosphatase conjugated with the secondary antibody. 1-Naphthol was determined by stopped flow injection analysis (SFIA) with electrochemical detector using a glassy carbon working electrode and by stationary electrochemical detection using linear sweep voltammetry. A special modular electrochemical SFIA system which needs only 10  μL of sample including working buffer for one analysis was completely designed and successfully verified. The recoveries in different matrices and analyte concentration were estimated. Detection limit (3 S/N) was estimated as 5  pg/mL of 8-OHdG. This method promises to be very easily modified to microfluidic systems as "lab on valve". The optimized method had sufficient selectivity and thus could be used for determination of 8-OHDG in human urine and therefore for estimation of oxidative DNA damage as a result of oxidation stress in prostate cancer patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31097
1-Naphthol, PESTANAL®, analytical standard
C10H8O