EMAIL THIS PAGE TO A FRIEND
Journal of virology
High-resolution profiling and analysis of viral and host small RNAs during human cytomegalovirus infection.
PMID 22013051
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-590
Anti-Ago 2 Antibody, clone, Upstate®, from rabbit