EMAIL THIS PAGE TO A FRIEND
Cephalalgia : an international journal of headache
Chronic daily cortical spreading depressions suppress spreading depression susceptibility.
PMID 22013142
Abstract
Migraine is a disabling chronic episodic disorder. Attack frequency progressively increases in some patients. Incremental cortical excitability has been implicated as a mechanism underlying progression. Cortical spreading depression (CSD) is the electrophysiological event underlying migraine aura, and a headache trigger. We hypothesized that CSD events during frequent migraine attacks condition the cortex to increase the susceptibility to further attacks. A single daily CSD was induced for 1 or 2 weeks in mouse frontal cortex; contralateral hemisphere served as sham control. At the end of CSD conditioning, occipital CSD susceptibility was determined by measuring the frequency of CSDs evoked by topical KCl application. Sham hemispheres developed 8.4 ± 0.3 CSDs/hour, and did not significantly differ from naïve controls without prior cranial surgery (9.3 ± 0.4 CSDs/hour). After 2 but not 1 week of daily CSD conditioning, CSD frequency (4.9 ± 0.3 CSDs/hour) as well as the duration and propagation speed were reduced significantly in the conditioned hemispheres. Histopathological examination revealed marked reactive astrocytosis without neuronal injury throughout the conditioned cortex after 2 weeks, temporally associated with CSD susceptibility. These data do not support the hypothesis that frequent migraine attacks predispose the brain to further attacks by enhancing tissue susceptibility to CSD.