EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Design, synthesis and biological evaluation of 3-substituted 2,5-dimethyl-N-(3-(1H-tetrazol-5-yl)phenyl)pyrroles as novel potential HIV-1 gp41 inhibitors.
PMID 22014749
Abstract
Based on the structure of HIV-1 gp41 binding site for small-molecule inhibitors, optimization of lead 2 resulted in the discovery of a new series of 2,5-dimethyl-3-(5-(N-phenylrhodaninyl)methylene)-N-(3-(1H-tetrazol-5-yl)phenyl)pyrrole compounds with improved anti-HIV-1 activity. The most active compounds 13a and 13j exhibited significant potency against gp41 6-HB formation with IC(50) values of 4.4 and 4.6 μM and against HIV-1 replication in the MT-2 cells with EC(50) values of 3.2 and 2.2 μM, respectively, thus providing a new starting point to develop highly potent small-molecule HIV fusion inhibitors targeting gp41.
Related Materials