EMAIL THIS PAGE TO A FRIEND
Respiratory medicine
Acetylsalicylic acid-triggered 15-HETE generation by peripheral leukocytes for identifying ASA sensitivity.
PMID 22015094
Abstract
Exposure to acetylsalicylic acid (ASA) may exacerbate respiratory or skin diseases or induce anaphylactoid reactions in apparently healthy individuals. We wanted to evaluate the clinical and diagnostic utility of measuring ASA-induced 15-hydroxyeicosatetraenoic acid (15-HETE) generation. We performed a prospective single-blind study with 26 subjects undergoing clinical evaluation and/or ASA provocation testing. We also included 12 control subjects. Peripheral blood leukocytes were incubated with 500 μM ASA and 15-HETE release was measured by competitive ELISA. We found that 18 subjects were ASA-tolerant and 8 were ASA-intolerant. The mean increase in 15-HETE in intolerant subjects was 34% and this was comparable to the mean increase of 30% observed in ASA-tolerant subjects. A similar mean increase was also observed in control subjects. The ROC calculation showed that the optimal diagnostic threshold would be an increase of greater than 33%. However, the sensitivity of this increase was only 63% and the specificity was 50%. Our data suggest that further studies are needed before the ASA-induced 15-HETE test can be used in clinical practice.