EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
BOP-mediated one-pot synthesis of C5-symmetric macrocyclic pyridone pentamers.
PMID 22016882
Abstract
We report here, for the first time, the BOP-mediated one-pot macrocyclization that is facilitated and guided by internally placed intramolecular H-bonds to allow for the highly selective formation of five-residue cation-binding macrocycles.