EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Reference intervals for orotic acid in urine, plasma and dried blood spot using hydrophilic interaction liquid chromatography-tandem mass spectrometry.
PMID 22019295
Abstract
Orotic acid (OA), a marker of hereditary orotic aciduria, is usually used for the differential diagnosis of some hyperammonemic inherited defects of urea cycle and of basic amino acid transporters. This study was aimed to establish age related reference intervals of OA in urine, and for the first time in plasma, and dried blood spot (DBS) from 229 apparently healthy subjects aged from three days to 40 years. The quantification of OA was performed by a previously implemented method, using a stable isotope dilution with 1,3-[(15)N(2)]-orotic acid and hydrophilic interaction liquid chromatography-tandem mass spectrometry (HILIC-MS/MS). The method has proved to be sensitive and accurate for a quantitative analysis of OA also in DBS and plasma. According to previous studies, urinary OA levels (mmol/mol of creatinine) decrease significantly with age. The upper limits (as 99th %ile) were of 3.44 and 1.30 in groups aged from three days to 1 year (group 1) and from 1 year to 12 years (group 2), respectively; in teenagers (from 13 to 19 years; group 3) and adults (from 20 to 40 years; group 4) urinary levels became more stable and the upper limits were of 0.64 and 1.21, respectively. Furthermore, OA levels in DBS (μM) also resulted significantly higher in subjects of group 1 (upper limit of 0.89) than in subjects of groups 2, 3 and 4 (upper limits of 0.24, 0.21, and 0.29, respectively). OA levels in plasma (μM) were significantly lower in subjects of group 3 (upper limit of 0.30) than in subjects of groups 1, 2, and 4 (upper limits of 0.59, 0.48, and 0.77, respectively). This method was also employed for OA quantification in plasma and DBS of 17 newborns affected by urea cycle defects, resulting sensitive and specific enough to screen these disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2750
Orotic acid, ≥98% (titration), anhydrous
C5H4N2O4