EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Monitoring the active transport of efflux pumps after their reconstitution into proteoliposomes: caveats and keys.
PMID 22019764
Abstract
There is an acute need for a functional assay allowing the investigation of efflux pumps. A dedicated procedure was previously developed, but although it was unambiguous, it suffered from a lack of reproducibility. We describe an optimization of the procedure that makes the assay much more robust.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60403
Potassium ionophore I, Selectophore, function tested
C54H90N6O18