EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Characterization, amyloid formation, and immobilization of a novel SGNH hydrolase from Listeria innocua 11262.
PMID 22020155
Abstract
A novel oligomeric hydrolase (LI22) from Listeria innocua CLIP 11262 was identified, characterized, and immobilized for industrial application. Sequence analysis of LI22 revealed a putative catalytic triad (Ser(10)-Asp(176)-His(179)), and a conserved sequence motif Ser(S)(10)-Gly(G)(77)-Asn(N)(79)-His(H)(179) with moderate identities (<30%) with other members of the SGNH-hydrolase superfamily. LI22 was able to hydrolyze p-nitrophenyl acetate, α- and β-naphthyl acetate, while the S10A mutant completely lost its activity. Structural properties of LI22 were investigated using gel filtration, circular dichroism (CD), fluorescence, molecular modeling, and gel filtration. We have shown that upon incubation in 30% TFE or 50% ethanol solution, LI22 was transformed into curly amyloid fibrils. Cross-linked enzyme aggregates of LI22 were prepared by precipitating the enzyme with ammonium sulfate and subsequent cross-linking with glutaraldehyde. Higher thermal and chemical stability, as well as good durability after repeated use of the LI22-CLEA, highlight its potential applicability as a biocatalyst in the pharmaceutical and chemical industries.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8130
4-Nitrophenyl acetate, esterase substrate
C8H7NO4
46021
4-Nitrophenyl acetate, ≥99.0% (GC)
C8H7NO4