EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Modification of histidyl residues in proteins by diethylpyrocarbonate.
PMID 22021
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40718
Diethyl pyrocarbonate, 99% (NT)
C6H10O5