EMAIL THIS PAGE TO A FRIEND
Tetrahedron
Sporolide B: Synthetic Studies.
PMID 22021939
Abstract
Studies directed towards the synthesis of the architecturally complex marine natural product sporolide B are described. Synthetic analysis suggested advanced hydroquinone and benzodiquinane fragments, which upon elaboration were successfully united via an ester linkage. Macrocyclization studies were then carried out, and although a novel macrocyclization product was obtained, subsequent studies revealed that the tertiary hydroxyls at C(6) and C(10) were sterically encumbered to participate in a successful macrocyclization to furnish sporolide B.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129127
1-Chloro-3,5-dibromobenzene, 95%
C6H3Br2Cl