EMAIL THIS PAGE TO A FRIEND
Metabolic engineering
Production of 3-hydroxypropionate homopolymer and poly(3-hydroxypropionate-co-4-hydroxybutyrate) copolymer by recombinant Escherichia coli.
PMID 22024131
Abstract
Conversion of 3-hydroxypropionate (3HP) from 1,3-propanediol (PDO) was improved by expressing dehydratase gene (dhaT) and aldehyde dehydrogenase gene (aldD) of Pseudomonas putida KT2442 under the promoter of phaCAB operon from Ralstonia eutropha H16. Expression of these genes in Aeromonas hydrophila 4AK4 produced up to 21 g/L 3HP in a fermentation process. To synthesize homopolymer poly(3-hydroxypropionate) (P3HP), and copolymer poly(3-hydroxypropionate-co-3-hydroxybutyrate) (P3HP4HB), dhaT and aldD were expressed in E. coli together with the phaC1 gene encoding polyhydroxyalkanoate (PHA) synthase gene of Ralstonia eutropha, and pcs' gene encoding the ACS domain of the tri-functional propionyl-CoA ligase (PCS) of Chloroflexus aurantiacus. Up to 92 wt% P3HP and 42 wt% P3HP4HB were produced by the recombinant Escherichia coli grown on PDO and a mixture of PDO+1,4-butanediol (BD), respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
493732
1,4-Butanediol, ReagentPlus®, 99%
C4H10O2
68891
1,4-Butanediol 4 M solution