EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Simultaneous determination of norfloxacin and lomefloxacin in milk by first derivative synchronous fluorescence spectrometry using Al (III) as an enhancer.
PMID 22027129
Abstract
A novel method for the simultaneous determination of norfloxacin (NFLX) and lomefloxacin (LFLX) in milk samples was developed by using first derivative synchronous fluorimetry. The synchronous fluorescence (Δλ=160 nm) spectra and first derivative synchronous fluorescence spectra of NFLX, LFLX and their mixture were studied. The zero-crossing method was utilized to measure the first derivative value of the derivative spectrum. The zero-crossing points were located at 275.0 nm for NFLX and at 283.8 nm for LFLX, in first derivative synchronous fluorescence spectra. Therefore, 283.8 nm and 275.0 nm were selected for the determination of NFLX and LFLX. The first derivative values varied linearly with the concentrations in the range of 1.68×10(-8)-5.64×10(-6) mol L(-1) for NFLX and 1.89×10(-8)-6.19×10(-6) mol L(-1) for LFLX. The detection limits were 5.03×10(-9) mol L(-1) for NFLX and 7.58×10(-9) mol L(-1) for LFLX. The proposed method is reliable, selective and sensitive, and has been used successfully in the simultaneous determination of NFLX and LFLX in milk samples, whose results were in good agreement with those obtained by HPLC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2906
Lomefloxacin hydrochloride
C17H19F2N3O3 · HCl