EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
X-ray computed tomography imaging of breast cancer by using targeted peptide-labeled bismuth sulfide nanoparticles.
PMID 22028313
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
333190
Bismuth(III) sulfide, 99%
Bi2S3