EMAIL THIS PAGE TO A FRIEND
Kinderkrankenschwester : Organ der Sektion Kinderkrankenpflege
["Of snakes and crocodiles": central side effects of nose drops and facts about rhinitis medicamentosa].
PMID 22029138
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6764
Triprolidine hydrochloride, ≥99%
C19H22N2 · HCl
X6000
Xylometazoline hydrochloride
C16H24N2 · HCl