EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Resonance Raman and density functional study of the excited state structural dynamics of 3-amino-2-cyclohexen-1-one in water and acetonitrile solvents.
PMID 22032972
Abstract
FT-Raman and/or FT-IR spectra of 3-amino-2-cyclohexen-1-one (ACyO) in solid state and/or in solvents of water and acetonitrile were obtained. Density functional theory calculations were done to help elucidate the vibrational band assignments. The A-band resonance Raman spectra of ACyO were acquired in water and acetonitrile solvents to examine the excited state structural dynamics and the state-mixing or curve-crossing tuned by solvents. A preliminary resonance Raman intensity analysis using the time-dependent wave-packet theory and simple model was done for ACyO in acetonitrile solvent. Resonance Raman spectroscopic probing of the excited state curve-crossing or state-mixing was proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102814
2-Cyclohexen-1-one, ≥95%
C6H8O