EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on 2-phenyl-2-propanol.
PMID 22033099
Abstract
A toxicologic and dermatologic review of 2-phenyl-2-propanol when used as a fragrance ingredient is presented. 2-Phenyl-2-propanol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a tertiary alcohol. The AAAs are a structurally diverse class of fragrance ingredients that includes primary, secondary, and tertiary alkyl alcohols covalently bonded to an aryl (Ar) group, which may be either a substituted or unsubstituted benzene ring. The common structural element for the AAA fragrance ingredients is an alcohol group -C-(R1)(R2)OH and generically the AAA fragrances can be represented as an Ar-C-(R1)(R2)OH or Ar-Alkyl-C-(R1)(R2)OH group. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. Available data for 2-phenyl-2-propanol were evaluated then summarized and includes physical properties, acute toxicity, skin irritation, skin sensitization, and toxicokinetics data. A safety assessment of the entire Aryl Alkyl Alcohols will be published simultaneously with this document; please refer to Belsito et al. (2012) for an overall assessment of the safe use of this material and all Aryl Alkyl Alcohols in fragrances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P30802
2-Phenyl-2-propanol, 97%
C9H12O