EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
A structural and spectroscopic study on para-aminohippuric acid with experimental and theoretical approaches.
PMID 22036005
Abstract
In this work, the molecular conformation, vibrational and electronic analysis of para-aminohippuric acid (pAHA, C(9)H(10)N(2)O(3)) were presented for the ground state using experimental techniques (FT-IR, FT-Raman and UV) and density functional theory (DFT) employing B3LYP exchange correlation with the 6-311++G(d,p) basis set. FT-IR and FT-Raman spectra were recorded in the regions of 400-4000cm(-1) and 50-4000cm(-1), respectively. The UV absorption spectra of the compound that dissolved in ethanol and water solution were recorded in the range of 190-400nm. Potential energy curve was computed by means of scanning NCCO torsion angle. The geometry optimization and the energies associated possible four conformers (C1-C4) were computed. The computational results diagnose the most stable conformer of pAHA as the C1 form. Optimized structure of compound was interpreted and compared with the earlier reported experimental values. The complete assignments of fundamental vibrations were performed on the basis of the total energy distribution (TED) of the vibrational modes, calculated with scaled quantum mechanics (SQM) method. A study on the electronic properties, such as frontier molecular energies, absorption wavelengths and oscillator strengths, were predicted by time-dependent DFT (TD-DFT) approach, while taking solvent effects into account. To investigate non-linear optical properties: polarizability, anisotropy of polarizability and molecular first hyperpolarizability of molecule were computed. Thermodynamic properties (heat capacity, entropy and enthalpy) of the title compound at different temperatures were calculated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1422
p-Aminohippuric acid, ≥99%
C9H10N2O3
08090
p-Aminohippuric acid, ≥98.0% (T)
C9H10N2O3