EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of deguelin-biotin conjugates and investigation into deguelin's interactions.
PMID 22037048
Abstract
Deguelin, a rotenoid, has emerged as an attractive pharmacophore for chemoprevention showing in vivo activity in several xenografts. Recently, several lines of evidence have suggested its mode of action may involve inhibition of HSP90, however binding in a different mode than known pharmacophores. To further probe the target of deguelin and related rotenoids, several biotin conjugates were prepared. None of the conjugates showed significant affinity for HSP90, however two conjugates showed a strong cellular co-localization with mitochondria, consistent with binding to mitochondrial complex 1. Contrarily to rotenone, deguelin and tephrosin were not found to inhibit tubulin polymerization demonstrating a dramatic pharmacological difference between these closely related rotenoids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0817
(−)-Deguelin, >98% (HPLC), powder
C23H22O6