EMAIL THIS PAGE TO A FRIEND
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
P-450-dependent epoxygenase pathway of arachidonic acid is involved in myeloma-induced angiogenesis of endothelial cells.
PMID 22038346
Abstract
P-450-dependent epoxygenase pathway of arachidonic acid and the products of epoxyeicosatrienoic acids (EETs) have been demonstrated to be involved in angiogenesis and tumor progression. This study examined the expression of EETs and the role of the pathway in the angiogenesis of multiple myeloma (MM). MM cell lines of U266 and RPMI8226 were cultured, and the EETs levels (11, 12-EET and 14, 15-EET) in the supernatant were determined by ELISA. Human umbilical vein endothelial cells (HUVECs) were cultured and used for analysis of the angiogenesis activity of the two MM cell lines, which was examined both in vitro and in vivo by employing MTT, chemotaxis, tube formation and matrigel plug assays. 11, 12-EET and 14, 15-EET were found in the supernatant of the cultured MM cells. The levels of the two EETs in the supernatant of U266 cells were significantly higher than those in the RPMI8226 cell supernatant (P<0.05), and the levels paralleled the respective angiogenesis activity of the two different MM cell lines. 17-octadecynoic acid (17-ODYA), as a specific inhibitor of P450 enzyme, suppressed HUVECs proliferation and tube formation induced by MM cells. Furthermore, 17-ODYA decreased the EET levels in the supernatant of MM cells. These results suggest that EETs may play an important role in the angiogenesis of MM, and the inhibitor 17-ODYA suppresses this effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O8382
17-Octadecynoic acid, ≥95% (GC)
C18H32O2