EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
The calcium-sensing receptor promotes adipocyte differentiation and adipogenesis through PPARγ pathway.
PMID 22038624
Abstract
Adipocyte differentiation and adipogenesis are closely related to obesity and obesity-induced metabolic disorders. The calcium-sensing receptor (CaSR) has been reported to play an antilipolytic role in human adipocyte and regulate cell differentiation in many tissues. However, the effects of CaSR on adipocyte differentiation and adipogenesis have not been clarified. In the study, we observed that activation of CaSR significantly promoted adipocyte differentiation and adipogenesis in human SW872 adipocytes. Gene expression analysis revealed that the CaSR activation increased the transcription factor proliferator-activated receptor γ (PPARγ) and its downstream genes including CCAAT element binding protein α (C/EBPα), adipose fatty acid-binding protein (aP2), and lipoprotein lipase. The activity of glycerol-3-phosphate dehydrogenase was also increased after the stimulation of CaSR. In addition, levels of cyclic AMP and calcium which have been shown to regulate PPARγ gene expression were significantly affected by the activation of CaSR. These effects could be suppressed by CaSR small interfering RNA (CaSR-siRNA). In conclusion, our findings suggest that activation of CaSR promotes differentiation and adipogenesis in adipocytes, which might be achieved by upregulating PPARγ and its downstream gene expressions. Therefore, CaSR in adipocytes may be involved in the pathogenesis of obesity by promoting adipocyte differentiation and adipogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
439770
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Cl3Gd