EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Aza-crown macrocycles as chiral solvating agents for mandelic acid derivatives.
PMID 22039861
Abstract
A series of new chiral macrocycles containing the trans-1,2-diaminocyclohexane (DACH) subunit and arene- and oligoethylene glycol-derived spacers has been prepared in enantiomerically pure form. Four of the macrocycles have been characterized by X-ray crystallography, which reveals a consistent mode of intramolecular N-H···N hydrogen bonding and conformational variations about the N-benzylic bonds. Most of the macrocycles were found to differentiate the enantiomers of mandelic acid (MA) by (1)H NMR spectroscopy in CDCl(3); within the series of macrocycles tested, enantiodiscrimination was promoted by (i) a meta-linkage geometry about the arene spacer, (ii) the presence of naphthalene- rather than phenylene-derived arene spacers, and (iii) increasing length of the oligoethylene glycol bridge. (1)H NMR titrations were performed with optically pure MA samples, and the data were fitted to a simultaneous 1:1 and 2:1 binding model, yielding estimates of 2:1 binding constants between some of the macrocycles and MA enantiomers. In several cases, NOESY spectra of the MA:macrocycle complexes show differential intramolecular correlations between protons adjacent to the amine and carboxylic acid groups of the macrocycles and MA enantiomers, respectively, thus demonstrating geometric differences between the diastereomeric intermolecular complexes. The three most effective macrocycles were employed as chiral solvating agents (CSAs) to determine the enantiomeric excess (ee) of 18 MA samples over a wide ee range and with very high accuracy (1% absolute error).