EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Functionalized 2,3-dihydrofurans via palladium-catalyzed oxyarylation of α-allyl-β-ketoesters.
PMID 22041862
Abstract
The palladium-catalyzed reaction of (hetero)aryl bromides, chlorides, and nonaflates with α-allyl-β-ketoesters provides ready efficient access to functionalized 2,3-dihydrofurans. The reaction tolerates several useful substituents including chloro, fluoro, ether, ketone, ester, cyano, and nitro groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
200018
2,3-Dihydrofuran, 99%
C4H6O