EMAIL THIS PAGE TO A FRIEND
Natural product research
A new diphenyl ether from Phoma sp. strain, SHZK-2.
PMID 22044095
Abstract
A new diphenyl ether methyl 2-(2-formyl-3-hydroxy-5-methylphenoxy)-5-hydroxy-3-methoxybenzoate (3), together with four known compounds, asterric acid (1), methyl asterrate (2), 9(Z),12(Z)-nonadecadienoic acid (4) and orsellinic acid (5), were isolated from the Phoma sp. strain SHZK-2, which was isolated from a polluted environment in southern China. The structures of these compounds were determined by spectroscopic methods. Cytotoxicities of compounds against HEPG2 cell and Raji cell lines were preliminarily evaluated by the MTT method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00012
Asterric acid, ≥95% (LC/MS-ELSD)
C17H16O8
SMB00570 Methyl asterrate, ≥95% (LC/MS-UV)
C18H18O8