EMAIL THIS PAGE TO A FRIEND
Natural product research
A new xanthone from Garcinia nitida.
PMID 22044165
Abstract
A detailed chemical study on the stem bark of Garcinia nitida has led to the isolation of five xanthones. They are 1,6-dihydroxy-5-methoxy-6,6-dimethylpyrano[2',3':2,3]-xanthone (1), inophyllin B (2), osajaxanthone (3), 3-isomangostin (4) and rubraxanthone (5). The structures of these compounds were established using mainly 1-D and 2-D NMR spectroscopy ((1)H, (13)C, DEPT, COSY, HMBC and HMQC) while molecular masses were determined via MS techniques; 1 is a new compound.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X600
Xanthone, 97%
C13H8O2