EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
Quorum sensing in fungi: Q&A.
PMID 22046125
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
79058
2-(4-Hydroxyphenyl)ethanol, analytical standard
C8H10O2