EMAIL THIS PAGE TO A FRIEND
Cell Death & Disease
Temporal blastemal cell gene expression analysis in the kidney reveals new Wnt and related signaling pathway genes to be essential for Wilms' tumor onset.
PMID 22048167