EMAIL THIS PAGE TO A FRIEND
Journal of periodontology
Effectiveness of systemic amoxicillin/metronidazole as an adjunctive therapy to full-mouth scaling and root planing in the treatment of aggressive periodontitis: a systematic review and meta-analysis.
PMID 22050545
Abstract
The systemic use of combined amoxicillin and metronidazole (AMX/MET) as an adjunctive treatment to full-mouth scaling and root planing (FMSRP) has been proposed for the treatment of generalized aggressive periodontitis; however, its effectiveness and clinical safety remain to be defined. The purpose of the present meta-analysis is to assess the effectiveness of FMSRP + AMX/MET compared to FMSRP alone. An electronic search of eight databases and a hand-search of 10 international dental journals were conducted through September 11, 2011. Gain in clinical attachment level (CAL), reduction in probing depth (PD), secondary outcomes, and adverse events were analyzed. A random-effect model was used to pool the extracted data. The weighted mean difference (MD) with 95% confidence interval (CI) was calculated for continuous outcomes, whereas risk difference (RD) with 95% CI was used for dichotomous data; heterogeneity was assessed with the χ(2)-based Cochran Q test and I(2) statistic. The level of significance was set at P <0.05. After the selection process, six randomized clinical trials were included. Results of the meta-analysis showed significant CAL gain (MD, 0.42; 95% CI, 0.23 to 0.61; P <0.05) and PD reduction (MD, 0.58; 95% CI, 0.39 to 0.77; P <0.05) in favor of FMSRP + AMX/MET; moreover, no significant RD was found in the occurrence of adverse events (RD, 0.01; 95% CI, -0.02 to 0.04; P >0.05). The findings of the meta-analysis seem to support the effectiveness and the clinical safety of FMSRP + AMX/MET; however, future studies are needed to confirm these results.