EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Fluorescent studies of salicylaldehyde and other related carbonyl compounds for the selective and sensitive detection of zinc(II) ions in aqueous solution.
PMID 22051980
Abstract
Salicylaldehyde was found to have a high selectivity for zinc ions with simultaneous enhancement of fluorescence in aqueous buffer solution at optimum pH 8.5. The stoichiometry of the complex was determined to be 1:1 with a K(a) value of 3.4 × 10(4) M(-1) at 298 K. The fluorescence of the complex is not affected by common anions and Zn(2+) binds preferentially to salicylaldehyde in the presence of alkali, alkaline earth and heavy metal cations (Hg(2+), Cd(2+), Cr(3+) and Ni(2+)). This property is not observed with related phenolic compounds bearing a carbonyl group such as esters, amides, carboxylic acids and ketones.