EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Homochirality beyond grinding: deracemizing chiral crystals by temperature gradient under boiling.
PMID 22053323
Abstract
A single-chirality solid phase can be obtained in boiling solutions containing a racemic mixture of left- and right-handed enantiomorphous crystals due to dissolution-crystallization cycles induced by a temperature gradient. This phenomenon provides further insights into asymmetric amplification mechanisms under presumably prebiotic conditions.