EMAIL THIS PAGE TO A FRIEND
Psychosomatics
"Ephedra-free" diet pill-induced psychosis.
PMID 22054631
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0752
(±)-Synephrine, ≥98%
C9H13NO2