EMAIL THIS PAGE TO A FRIEND
Glycobiology
Sialome analysis of the cephalochordate Branchiostoma belcheri, a key organism for vertebrate evolution.
PMID 22057003
Abstract
Sialic acid, a common terminal substitution of glycoconjugates, has been so far consistently identified in all vertebrates as well as in a growing number of bacterial species. It is assumed to be widely distributed among animal species of the deuterostome phylum, based on its identification in few echinoderm and all vertebrate species. However, whole sections of deuterostome, especially those intermediate species between invertebrates and vertebrates including cephalochordates, urochordates and hemichordates, are still unexplored in term of sialylation capacities. The discovery of functional sialic acid machinery in some of these species may shed new light onto the evolution of glycosylation capacities in deuterostome lineage. In a first approach, we investigated the sialylation pattern of a cephalocordate species, Branchiostoma belcheri, which occupies a strategic phylogenetic position to understand the transition of invertebrates toward vertebrates. Structural analysis of B. belcheri glycoconjugates established that this organism synthesizes large quantities of various sialic acids, some of which present rare or novel structures such as methylated sialic acids. These sialic acids were shown to be mainly associated with mono- and disialylated core 1-type O-glycans. Moreover, screening of the animal organs revealed the existence of exquisite tissue specificity in the distribution of sialic acids. Description of sialylation profiles was then correlated with the expression patterns of key enzymes involved in the biosynthesis of major forms of sialic acids, which provides the first complete overview of the sialylation patterns in cephalochordates.