EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ratiometric fluorescence imaging for distinguishing chloride concentration between normal and ischemic ventricular myocytes.
PMID 22057128
Abstract
We devised a new ratiometric fluorescent probe for the detection of chloride ions. This synthesized probe was applied to the ventricular myocytes to successfully realize dynamic imaging of Cl(-) concentration fluctuations during the myocardial ischemia course.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183067
6-Methoxyquinoline, 98%
C10H9NO
201626
Fluoresceinamine, isomer I
C20H13NO5