EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
n-Doping of organic electronic materials using air-stable organometallics.
PMID 22057596
Abstract
Air-stable dimers of sandwich compounds including rhodocene and (pentamethylcyclopentadienyl)(arene)ruthenium and iron derivatives can be used for n-doping electron-transport materials with electron affinities as small as 2.8 eV. A p-i-n homojunction diode based on copper phthalocyanine and using rhodocene dimer as n-dopant shows a rectification ratio of greater than 10(6) at 4 V.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
795615
Pentamethylcyclopentadienyl cyclopentadienyl rhodium dimer
C30H40Rh2