EMAIL THIS PAGE TO A FRIEND
Human Molecular Genetics
Derivation of new human embryonic stem cell lines reveals rapid epigenetic progression in vitro that can be prevented by chemical modification of chromatin.
PMID 22058289