EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Selective molecular sequestration with concurrent natural product functionalization and derivatization: from crude natural product extracts to a single natural product derivative in one step.
PMID 22059469
Abstract
A resin-bound nitroso compound sequestered a single unexpected component from crude plant seed extracts. Several plants, including Piper nigrum, Eugenia caryophyllata, and Pimenta dioica, were extracted with organic solvent in the presence of a nitroso-containing resin. The nitroso resin selectively sequestered a single compound, β-caryophyllene, via a chemo- and regioselective ene reaction. The ene product was released from the resin, and proper selection of the solid-phase linker and cleavage cocktail allowed concomitant further transformation of the primary ene product to a novel functionalized polycycle. Preliminary studies indicate that the new hydroxylamine-containing natural product derivatives have antibiotic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28910
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
C6H8
125415
1,4-Cyclohexadiene, 97%
C6H8