EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Iridium-catalyzed allylic fluorination of trichloroacetimidates.
PMID 22059470
Abstract
A rapid allylic fluorination method utilizing trichloroacetimidates in conjunction with an iridium catalyst has been developed. The reaction is conducted at room temperature under ambient air and relies on Et(3)N·3HF reagent to provide branched allylic fluorides with complete regioselectivity. This high-yielding reaction can be conducted on a multigram scale and shows considerable functional group tolerance. The use of [(18)F]KF·Kryptofix allowed (18)F(-) incorporation in 10 min.