EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Inhibition of glutaminyl cyclase attenuates cell migration modulated by monocyte chemoattractant proteins.
PMID 22060158
Abstract
QC (glutaminyl cyclase) catalyses the formation of N-terminal pGlu (pyroglutamate) in peptides and proteins. pGlu formation in chemoattractants may participate in the regulation of macrophage activation and migration. However, a clear molecular mechanism for the regulation is lacking. The present study examines the role of QC-mediated pGlu formation on MCPs (monocyte chemoattractant proteins) in inflammation. We demonstrated in vitro the pGlu formation on MCPs by QC using MS. A potent QC inhibitor, PBD150, significantly reduced the N-terminal uncyclized-MCP-stimulated monocyte migration, whereas pGlu-containing MCP-induced cell migration was unaffected. QC small interfering RNA revealed a similar inhibitory effect. Lastly, we demonstrated that inhibiting QC can attenuate cell migration by lipopolysaccharide. These results strongly suggest that QC-catalysed N-terminal pGlu formation of MCPs is required for monocyte migration and provide new insights into the role of QC in the inflammation process. Our results also suggest that QC could be a drug target for some inflammatory disorders.