EMAIL THIS PAGE TO A FRIEND
Natural product research
Phytochemical study of essential oil from the aerial parts of Coleus aromaticus Benth.
PMID 22060194
Abstract
The essential oil composition of Coleus aromaticus Benth. (family Lamiaceae) was examined by capillary GC and GC-MS. Analyses revealed the presence of 28 constituents, of which 16 were identified. Thymol (83.39%) was found to be the major compound, while 1-octen-3-ol, terpine-4-ol, eugenol, trans-caryophyllene, caryophyllene oxide and α-cadinol were present as minor constituents.