EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Carboxylate-directed tandem functionalisations of α,β-dihaloalkenoic acids with 1-alkynes: a straightforward access to (Z)-configured, α,β-substituted γ-alkylidenebutenolides.
PMID 22065483
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283754
2(5H)-Furanone, 98%
C4H4O2