EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Effects of isopsoralen on bone marrow stromal stem cells differentiate and proliferate in vitro].
PMID 22066454
Abstract
To investigated the effects of isopsoralen on bone marrow stromal stem cells (BMSCs) differentiate and proliferate in vitro. The stratum of mononuclear cells were separated and collected from the rat bone marrow sample by the all bone marrow cell culture methods. The cells were cultured in DMEM contained 10% fetal bovine serum. The culture medium was changed after three days. Nine days later, cells were treatment by isopsoralen with the concentration 1 x 10(-5), 1 x 10(-4), 1 x10(-6), 1 x 10(-7) mol x L(-1). MTT method was used for the proliferated analyzing. Under the induced condition, the alkaline phosphatase (ALP) activity, calcium salt sediment yield and osteocalcin were measured at the 4, 8, 12, 16 d. At the fifteenth day, histochemistry dyeing for calcified tubercle and ALP was proceeded. Total RNA was isolated and the gene expression of bFGF, IGF-1, Osterix and Runx-2 were investigated by Real Time PCR. The BMSCs proliferation refrained by isopsoralen with dose dependent. But it significantly enhanced osteogenesis, which was represented by the promotion of the ALP activity, calcium salt sediment yield, osteocalcin, and calcified tubercle amount. Besides, it also enhanced the mRNA level of bFGF, IGF-1, Osterix and Runx-2. The isopsoralen with the concentration 1 x 10(-5) mol x L(-1) can promote BMSCs differentiation to osteoblasts. It demonstrated the isopsoralen can prevent antiosteoporotic, which is an active part of the traditional Chinese medicine psoralea corylifolia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0956
Angelicin
C11H6O3