EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Nickel-catalyzed Heck-type reactions of benzyl chlorides and simple olefins.
PMID 22066899
Abstract
Nickel-catalyzed intermolecular benzylation and heterobenzylation of unactivated alkenes to provide functionalized allylbenzene derivatives are described. A wide range of both the benzyl chloride and alkene coupling partners are tolerated. In contrast to analogous palladium-catalyzed variants of this process, all reactions described herein employ electronically unbiased aliphatic olefins (including ethylene), proceed at room temperature, and provide 1,1-disubstituted olefins over the more commonly observed 1,2-disubstituted olefins with very high selectivity.
Related Materials