EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
(AEDPH3)·(BtaH): a novel supramolecular plaster with formaldehyde adsorption and formaldehyde/ultraviolet ray-induced luminescence switching performance.
PMID 22068261
Abstract
A novel supramolecular plaster, (AEDPH(3))·(BtaH) (1), is synthesised and characterized. The supramolecular plaster is easy to synthesise and process, and displays good mechanical properties. It can adsorb and eliminate formaldehyde (HCHO) with high efficiency and exhibits very interesting HCHO/ultraviolet ray-induced luminescence switching.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
268674
2-Aminoethylphosphonic acid, 99%
C2H8NO3P