EMAIL THIS PAGE TO A FRIEND
Brain and nerve = Shinkei kenkyu no shinpo
[Neuroimaging Findings of a 29-year-old woman with 22q11.2 deletion presenting with cognitive deterioration].
PMID 22068483
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I120
Iofetamine hydrochloride, analytical standard, for drug analysis
C12H18IN · HCl