EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Kinetics of ergothioneine inhibition of mushroom tyrosinase.
PMID 22068690
Abstract
The native amino acid ergothioneine, a thiourea derivative of histidine, inhibits mushroom tyrosinase activity in a dose-dependent manner, with an IC(50) value of 1.025 mg/ml (4.47 mM). By contrast, histidine exhibited no inhibitory effect on mushroom tyrosinase activity. We characterized ergothioneine as a noncompetitive tyrosinase inhibitor using a Lineweaver-Burk plot of experimental kinetic data. The IC(50) value for ergothioneine scavenging of 2,2-diphenyl-1-picrylhydrazyl was 6.110 ± 0.305 mg/ml, much higher than the IC(50) for inhibition of tyrosinase activity which indicating ergothioneine on tyrosinase shows a weak correlation to its antioxidative activity. The results demonstrated that ergothioneine has a potent inhibition effect on tyrosinase enzyme activity, resulting from the presence of the sulfur substituted imidazole ring in ergothioneine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7521
L-(+)-Ergothioneine
C9H15N3O2S