EMAIL THIS PAGE TO A FRIEND
Anais brasileiros de dermatologia
Pyodermatitis-pyostomatitis vegetans: case report and review of medical literature.
PMID 22068794
Abstract
Pyodermatitis-pyostomatitis vegetans is a rare inflammatory dermatosis of unknown etiology, with a typical mucocutaneous involvement. We report the case of a woman with pustular and vesicular lesions in the axillae, evolving with vegetating plaques and pustules with annular grouping. The disease progressed with vulvar and inguinal involvement as well as involvement of the oral, nasal and ocular mucous membranes. She started the treatment with prednisone (40 mg/day), with remission of the lesions after one month of use of such medication. Association with inflammatory bowel disease occurs in 70% of the cases. Immunofluorescence, which is typically negative, helps to characterize the disease. A rapid response to systemic steroids is expected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4520
Cephalothin sodium salt
C16H15N2NaO6S2