EMAIL THIS PAGE TO A FRIEND
Toxins
Effect of antioxidant mixtures on growth and ochratoxin a production of Aspergillus section Nigri species under different water activity conditions on peanut meal extract agar.
PMID 22069644
Abstract
The effect of mixtures of antioxidants butylated hydroxyanisol (BHA) and propyl paraben (PP) on lag phase, growth rate and ochratoxin A (OTA) production by four Aspergillus section Nigri strains was evaluated on peanut meal extract agar (PMEA) under different water activities (a(w)). The antioxidant mixtures used were: BHA + PP (mM), M1 (0.5 + 0.5), M2 (1.0 + 0.5), M3 (2.5 + 0.5), M4 (0.5 + 1.0), M5 (1.0 + 1.0), M6 (2.5 + 1.0), M7 (5.0 + 2.5) and M8 (10 + 2.5). The mixture M8 completely suppressed mycelial growth for all strains. A significant stimulation in OTA production was observed with mixtures M1 to M5 mainly at the highest a(w); whereas M6, M7 and M8 completely inhibited OTA production in all strains assayed; except M6 in A. carbonarius strain (RCP G). These results could enable a future intervention strategy to minimize OTA contamination.