EMAIL THIS PAGE TO A FRIEND
Toxins
Development and characterization of a monoclonal antibody against Ochratoxin B and its application in ELISA.
PMID 22069651
Abstract
A monoclonal antibody specific to ochratoxin B (OTB) was employed for the development of an indirect competitive OTB-ELISA. The optimized OTB-ELISA resulted in a limit of detection (LOD) for OTB of 3 µg/L (8 nM), a limit of quantification (LOQ) of 3.7 µg/L (10 nM), and a 50% inhibitory concentration (IC(50)) of 150 nM. Due to very low cross-reactivity to OTA (2.7%) and structurally related molecules (0%), this OTB-ELISA was found to be suitable to detect OTB with excellent precision in different matrices, i.e., beer, coffee and wine. Therefore, this OTB-ELISA will allow screening of OTB in food and feed products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32411
Ochratoxin B solution, 10 μg/mL in acetonitrile, analytical standard
C20H19NO6